siRNA Details
Gene Symbol: Arap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229897.2 | XP_006229959.1 | 17 | 2599 | ||
XM_006229898.2 | XP_006229960.1 | 17 | 3343 | |||
XM_006229899.2 | XP_006229961.1 | 17 | 3341 | |||
XM_006229900.1 | XP_006229962.1 | 15 | 2218 | |||
XM_006229901.1 | XP_006229963.1 | 14 | 2039 | |||
XM_008759810.1 | XP_008758032.1 | 18 | 3766 | |||
XM_008759811.1 | XP_008758033.1 | 17 | 3341 |
