siRNA Details
Gene Symbol: Stim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108496.2 | NP_001101966.2 | 11, 12 | 1915 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009287, U47323, AK201190, BC021644, AK041944.
Gene Ontology Categories:
