siRNA Details
Gene Symbol: Stim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108496.2 | NP_001101966.2 | 1 | 465 | ||
GenBank mRNA | BC166914.1 | - | 1 | 124 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108496.2 | NP_001101966.2 | 1 | 465 | ||
GenBank mRNA | BC166914.1 | - | 1 | 124 |