siRNA Details
Gene Symbol: Hbb-b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198776.1 | NP_942071.1 | 2 | 167 | ||
XM_006229913.2 | XP_006229975.2 | 2 | 201 | |||
GenBank mRNA | BC058502.1 | - | 2 | 167 | 6887460 | |
FQ218177.1 | - | 2 | 161 |
Gene Symbol: LOC100134871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113223.1 | NP_001106694.1 | 2 | 162 | ||
GenBank mRNA | FQ209951.1 | - | 2 | 165 | ||
FQ210023.1 | - | 2 | 165 | |||
FQ210060.1 | - | 2 | 163 | |||
FQ210106.1 | - | 2 | 163 | |||
FQ210483.1 | - | 2 | 163 | |||
FQ210600.1 | - | 2 | 165 | |||
FQ210669.1 | - | 2 | 159 | |||
FQ210740.1 | - | 2 | 163 | |||
FQ211266.1 | - | 2 | 163 | |||
FQ211272.1 | - | 2 | 163 | |||
FQ211275.1 | - | 2 | 163 | |||
FQ211294.1 | - | 2 | 165 | |||
FQ211362.1 | - | 2 | 163 | |||
FQ211415.1 | - | 2 | 163 | |||
FQ211424.1 | - | 2 | 163 | |||
FQ211697.1 | - | 2 | 165 | |||
FQ212253.1 | - | 2 | 163 | |||
FQ212317.1 | - | 2 | 165 | |||
FQ212406.1 | - | 2 | 165 | |||
FQ212421.1 | - | 2 | 163 | |||
FQ213080.1 | - | 2 | 192 | |||
FQ214057.1 | - | 2 | 163 | |||
FQ214129.1 | - | 2 | 163 | |||
FQ217131.1 | - | 2 | 161 | |||
FQ217673.1 | - | 2 | 163 | |||
FQ218002.1 | - | 2 | 164 | |||
FQ218191.1 | - | 2 | 163 | |||
FQ218246.1 | - | 2 | 135 | |||
FQ218266.1 | - | 2 | 201 | |||
FQ218689.1 | - | 2 | 163 | |||
FQ218821.1 | - | 2 | 163 | |||
FQ219109.1 | - | 2 | 163 | |||
FQ219133.1 | - | 2 | 194 | |||
FQ221588.1 | - | 2 | 161 | |||
FQ222844.1 | - | 2 | 163 | |||
FQ224149.1 | - | 2 | 163 | |||
FQ224301.1 | - | 2 | 163 | |||
FQ224598.1 | - | 2 | 163 | |||
FQ224632.1 | - | 2 | 163 | |||
FQ224738.1 | - | 2 | 163 | |||
FQ224763.1 | - | 2 | 202 | |||
FQ225377.1 | - | 2 | 163 | |||
FQ225739.1 | - | 2 | 165 | |||
FQ226460.1 | - | 2 | 163 | |||
FQ226535.1 | - | 2 | 163 | |||
FQ226609.1 | - | 2 | 165 | |||
FQ226955.1 | - | 2 | 163 | |||
FQ228555.1 | - | 2 | 163 | |||
FQ230881.1 | - | 2 | 165 | |||
FQ232298.1 | - | 307 | ||||
FQ232442.1 | - | 2 | 194 | |||
FQ233001.1 | - | 2 | 163 | |||
FQ234495.1 | - | 2 | 164 | |||
X05080.1 | - | 2 | 162 | |||
X16418.1 | - | 2 | 128 |
Gene Symbol: LOC103694855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773799.1 | XP_008772021.1 | 2 | 203 | ||
XM_008773800.1 | XP_008772022.1 | 2 | 197 |
Gene Symbol: LOC103694857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773801.1 | XP_008772023.1 | 2 | 199 |
Gene Symbol: LOC689064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111269.1 | NP_001104739.1 | 145 | |||
GenBank mRNA | DY319996.1 | - | 145 | |||
FQ209804.1 | - | 165 | ||||
FQ209902.1 | - | 165 | ||||
FQ210004.1 | - | 162 | ||||
FQ210039.1 | - | 165 | ||||
FQ210298.1 | - | 163 | ||||
FQ210379.1 | - | 165 | ||||
FQ210599.1 | - | 163 | ||||
FQ210742.1 | - | 163 | ||||
FQ210889.1 | - | 165 | ||||
FQ211252.1 | - | 163 | ||||
FQ211410.1 | - | 163 | ||||
FQ211471.1 | - | 162 | ||||
FQ211709.1 | - | 238 | ||||
FQ215055.1 | - | 162 | ||||
FQ217121.1 | - | 194 | ||||
FQ217477.1 | - | 163 | ||||
FQ217746.1 | - | 163 | ||||
FQ218196.1 | - | 163 | ||||
FQ218287.1 | - | 163 | ||||
FQ218323.1 | - | 163 | ||||
FQ221211.1 | - | 163 | ||||
FQ225430.1 | - | 163 | ||||
FQ225765.1 | - | 163 | ||||
FQ226110.1 | - | 163 | ||||
FQ228130.1 | - | 693 | ||||
FQ230470.1 | - | 165 | ||||
FQ230522.1 | - | 170 | ||||
FQ230899.1 | - | 167 | ||||
M27170.1 | - | 21 | ||||
M27171.1 | - | 21 | ||||
M27173.1 | - | 21 |

More Information
Other Species Targeted

Gene Ontology Categories:
