siRNA Details
Gene Symbol: Trim66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191839.1 | NP_001178768.1 | 12 | 2726 | ||
XM_006229983.2 | XP_006230045.1 | 13 | 3923 | |||
GenBank mRNA | BC168212.1 | - | 13 | 2613 |
