siRNA Details
Gene Symbol: Akip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108497.1 | NP_001101967.1 | 5 | 801 | ||
XM_006229987.1 | XP_006230049.1 | 6 | 690 | |||
XM_006229988.2 | XP_006230050.1 | 6 | 621 | |||
XM_006229989.1 | XP_006230051.1 | 6 | 660 | |||
XM_006229990.1 | XP_006230052.1 | 6 | 648 | |||
XM_006229991.1 | XP_006230053.1 | 5 | 609 | |||
XM_006229992.1 | XP_006230054.1 | 5 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
