siRNA Details
Gene Symbol: Tead1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198589.1 | NP_001185518.1 | 10 | 1058 | ||
XM_006230052.2 | XP_006230114.1 | 11 | 1403 | |||
XM_006230053.2 | XP_006230115.1 | 10 | 1391 | |||
XM_006230054.2 | XP_006230116.1 | 10 | 1340 | |||
XM_006230055.2 | XP_006230117.1 | 10 | 1340 | |||
XM_006230056.2 | XP_006230118.1 | 9 | 1328 | |||
XM_006230057.2 | XP_006230119.1 | 9 | 1328 | |||
XM_006230058.2 | XP_006230120.1 | 8 | 1265 | |||
XM_006230059.2 | XP_006230121.1 | 11 | 2147 | |||
XM_008759767.1 | XP_008757989.1 | 11 | 1597 | |||
XM_008759768.1 | XP_008757990.1 | 11 | 960 | |||
GenBank mRNA | AY529194.1 | - | 9 | 788 | ||
AY529195.1 | - | 9 | 788 | |||
AY529196.1 | - | 8 | 776 | |||
AY529197.1 | - | 8 | 725 | |||
AY529198.1 | - | 7 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
