siRNA Details
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108502.2 | NP_001101972.2 | 9 | 1271 | ||
XM_006230127.2 | XP_006230189.1 | 10 | 1424 | |||
XM_006230129.2 | XP_006230191.1 | 9 | 1187 | |||
XM_008759771.1 | XP_008757993.1 | 10 | 1340 | |||
XM_008759772.1 | XP_008757994.1 | 9 | 1316 | |||
XM_008759773.1 | XP_008757995.1 | 10 | 1421 | |||
GenBank mRNA | BC168985.1 | - | 9 | 1271 | 7323876 | |
FQ218416.1 | - | 5 | 464 |
