siRNA Details
Gene Symbol: Polr3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108503.1 | NP_001101973.1 | 13 | 1124 | ||
XM_006230168.2 | XP_006230230.1 | 13 | 1153 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Polr3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108503.1 | NP_001101973.1 | 13 | 1124 | ||
XM_006230168.2 | XP_006230230.1 | 13 | 1153 |