siRNA Details
Gene Symbol: Spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039208.2 | NP_001034297.2 | 2 | 615 | ||
GenBank mRNA | BC110048.1 | - | 2 | 600 | 7315879 | |
CA339235.1 | - | 2 | 188 | |||
CK604160.1 | - | 2 | 615 |

More Information
Other Species Targeted

Gene Ontology Categories:
