siRNA Details
Gene Symbol: RGD1310127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035517.1 | NP_001030594.1 | 11, 12 | 1262 | ||
GenBank mRNA | BC099813.1 | - | 11, 12 | 1262 | 7930179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
