siRNA Details
Gene Symbol: Tial1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013193.1 | NP_001013211.1 | 11, 12 | 1255 | ||
XM_006230310.2 | XP_006230372.1 | 11, 12 | 1461 | |||
XM_008759902.1 | XP_008758124.1 | 11, 12 | 1280 | |||
GenBank mRNA | BC091409.1 | - | 11, 12 | 1255 | 7325481 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009383, U55861, AK012085, BC068194, BC010496, and others.
Gene Ontology Categories:
