siRNA Details
Gene Symbol: Btbd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017464.2 | NP_001017464.1 | 14 | 1423 | ||
XM_006230403.2 | XP_006230465.1 | 13 | 1436 | |||
XM_008759903.1 | XP_008758125.1 | 14 | 1523 | |||
XM_008759904.1 | XP_008758126.1 | 12 | 1189 | |||
XM_008759905.1 | XP_008758127.1 | 14 | 1523 | |||
XM_008759906.1 | XP_008758128.1 | 14 | 1519 | |||
GenBank mRNA | BC079317.1 | - | 14 | 1426 | 7135047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
