siRNA Details
Gene Symbol: SUCLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177599.1 | NP_001171070.1 | 2, 3 | 251 | ||
NM_003848.3 | NP_003839.2 | 2, 3 | 251 | |||
XM_017007415.1 | XP_016862904.1 | 2, 3 | 247 | |||
XM_017007416.1 | XP_016862905.1 | 2, 3 | 247 | |||
XM_017007417.1 | XP_016862906.1 | 2, 3 | 247 | |||
XM_017007418.1 | XP_016862907.1 | 2, 3 | 247 | |||
XM_017007419.1 | XP_016862908.1 | 2, 3 | 247 | |||
XM_017007420.1 | XP_016862909.1 | 2, 3 | 247 | |||
GenBank mRNA | AF058954.1 | - | 1, 2 | 139 | ||
AK310527.1 | - | 2, 3 | 247 | |||
BC007716.2 | - | 2, 3 | 214 | 4303307 | ||
BC035149.1 | - | 1, 2 | 13 | |||
BC047024.1 | - | 3, 4 | 221 | 5415743 | ||
BC068602.1 | - | 2, 3 | 252 | 5274341 |
