siRNA Details
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108505.2 | NP_001101975.2 | 5 | 559 | ||
GenBank mRNA | FQ227185.1 | - | 5 | 550 | ||
FQ228402.1 | - | 5 | 557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
