siRNA Details
Gene Symbol: Ifitm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136124.1 | NP_001129596.1 | 1 | 173 | ||
GenBank mRNA | AF164039.1 | - | 1 | 150 | ||
BC168774.1 | - | 1 | 173 | 6921753 |

More Information
Other Species Targeted

Gene Ontology Categories:
