siRNA Details
Gene Symbol: Tollip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109668.1 | NP_001103138.1 | 6 | 1476 | ||
XM_006230607.2 | XP_006230669.1 | 7 | 1557 | |||
XM_008760003.1 | XP_008758225.1 | 6 | 1421 | |||
GenBank mRNA | BC133067.1 | - | 6 | 1362 | 7193007 |

More Information
Other Species Targeted

Gene Ontology Categories:
