siRNA Details
Gene Symbol: Lsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025420.1 | NP_001020591.1 | 3, 4 | 423 | ||
XM_006230815.2 | XP_006230877.1 | 4, 5 | 1085 | |||
XM_006230817.2 | XP_006230879.1 | 3, 4 | 460 | |||
XM_006230818.1 | XP_006230880.1 | 3, 4 | 518 | |||
XM_008760146.1 | XP_008758368.1 | 3, 4 | 498 | |||
XM_008760147.1 | XP_008758369.1 | 3, 4 | 747 | |||
GenBank mRNA | BC097966.1 | - | 3, 4 | 397 | 7388019 | |
CO556739.1 | - | 3, 4 | 422 | |||
FQ230230.1 | - | 3, 4 | 404 | |||
FQ232610.1 | - | 3, 4 | 413 | |||
FQ233963.1 | - | 3, 4 | 413 |
