siRNA Details
Gene Symbol: Osbpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015024.2 | NP_001015024.2 | 21 | 2747 | ||
XM_006230828.2 | XP_006230890.1 | 21 | 2555 | |||
XM_006230829.2 | XP_006230891.1 | 21 | 2555 | |||
XM_006230830.2 | XP_006230892.1 | 21 | 2641 | |||
GenBank mRNA | BC091326.1 | - | 4 | 669 |
