siRNA Details
Gene Symbol: LOC102555167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760197.1 | XP_008758419.1 | 1 | 259 |
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109979.2 | NP_001103449.1 | 291 | |||
GenBank mRNA | BC085118.1 | - | 258 | |||
DN934711.1 | - | 309 | ||||
EV762710.1 | - | 310 |

More Information
Other Species Targeted

Gene Ontology Categories:
