siRNA Details
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109979.2 | NP_001103449.1 | 240 | |||
GenBank mRNA | BC085118.1 | - | 207 | |||
DN934711.1 | - | 258 | ||||
EV762710.1 | - | 259 | ||||
X13905.1 | - | 180 |
Gene Symbol: LOC102555167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760197.1 | XP_008758419.1 | 1 | 208 |
