siRNA Details
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004089.1 | NP_001004089.1 | 2 | 779 | ||
XM_006230849.2 | XP_006230911.1 | 3 | 1396 | |||
GenBank mRNA | AB091474.1 | - | 2 | 779 | ||
FQ223350.1 | - | 3 | 986 | |||
FQ228273.1 | - | 3 | 965 |
