siRNA Details
Gene Symbol: Sipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004089.1 | NP_001004089.1 | 9, 10 | 2524 | ||
XM_006230849.2 | XP_006230911.1 | 10, 11 | 3141 | |||
GenBank mRNA | AB091474.1 | - | 9, 10 | 2524 | ||
FQ228273.1 | - | 10, 11 | 2710 |
