siRNA Details
Gene Symbol: Rasgrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082977.1 | NP_001076446.1 | 6 | 607 | ||
XM_006230853.2 | XP_006230915.1 | 6 | 635 | |||
XM_006230854.2 | XP_006230916.1 | 6 | 1061 | |||
XM_006230855.2 | XP_006230917.1 | 6 | 1102 | |||
XM_006230857.2 | XP_006230919.1 | 6 | 630 | |||
XM_008760158.1 | XP_008758380.1 | 6 | 781 |

More Information
Other Species Targeted

Gene Ontology Categories:
