siRNA Details
Gene Symbol: RGD1308106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134575.1 | NP_001128047.1 | 5 | 927 | ||
XM_006230863.2 | XP_006230925.1 | 5 | 1135 | |||
XM_006230864.2 | XP_006230926.1 | 5 | 1135 |
