siRNA Details
Gene Symbol: RGD1308106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134575.1 | NP_001128047.1 | 5 | 923 | ||
XM_006230863.2 | XP_006230925.1 | 5 | 1131 | |||
XM_006230864.2 | XP_006230926.1 | 5 | 1131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
