siRNA Details
Gene Symbol: Scgb2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203333.2 | NP_976078.1 | 2 | 234 | ||
XM_008760232.1 | XP_008758454.1 | 2 | 533 | |||
GenBank mRNA | BC062401.1 | - | 2 | 194 | 5621455 | |
CB791758.1 | - | 2 | 246 | |||
J00772.1 | - | 2 | 185 | |||
V01263.1 | - | 2 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
