siRNA Details
Gene Symbol: Dak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039031.1 | NP_001034120.1 | 5 | 754 | ||
XM_006231056.2 | XP_006231118.1 | 5 | 640 | |||
XM_006231057.2 | XP_006231119.2 | 5 | 923 | |||
GenBank mRNA | BC098925.1 | - | 5 | 754 | 7112371 |
