siRNA Details
Gene Symbol: Ms4a4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760273.1 | XP_008758495.1 | 4 | 3024 | ||
XM_342027.5 | XP_342028.3 | 3 | 4639 |
Gene Symbol: LOC100911403 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231085.2 | XP_006231147.1 | 2 | 327 |
