siRNA Details
Gene Symbol: Patl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108520.2 | NP_001101990.1 | 10 | 1277 | ||
GenBank mRNA | BC168974.1 | - | 10 | 1277 | 7106310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
