siRNA Details
Gene Symbol: Patl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108520.2 | NP_001101990.1 | 10 | 1307 | ||
GenBank mRNA | BC168974.1 | - | 10 | 1307 | 7106310 |

More Information
Other Species Targeted

Gene Ontology Categories:
