siRNA Details
Gene Symbol: RGD1359158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007737.1 | NP_001007738.1 | 3, 4 | 371 | ||
GenBank mRNA | BC079346.1 | - | 3, 4 | 371 | 7107242 | |
BC093390.1 | - | 3, 4 | 362 | 7388009 | ||
FQ215421.1 | - | 2, 3 | 177 |
