siRNA Details
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014168.1 | NP_001014190.1 | 2 | 118 | ||
GenBank mRNA | BC085841.1 | - | 2 | 118 | 7135812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
