siRNA Details
Gene Symbol: Sfxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108527.1 | NP_001101997.1 | 7, 8 | 446 | ||
XM_006231684.2 | XP_006231746.1 | 6, 7 | 2507 | |||
XM_006231685.2 | XP_006231747.1 | 6, 7 | 1343 | |||
XM_008760590.1 | XP_008758812.1 | 5, 6 | 2241 | |||
GenBank mRNA | FQ215895.1 | - | 7, 8 | 435 |
