siRNA Details
Gene Symbol: Zfp57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213565.4 | NP_998730.2 | 5 | 1210 | ||
XM_008772694.1 | XP_008770916.1 | 4 | 1383 | |||
GenBank mRNA | BC127468.1 | - | 3 | 1028 | 8368087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
