siRNA Details
Gene Symbol: RT1-T24-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008858.1 | NP_001008858.1 | 931 | |||
XM_008772730.1 | XP_008770952.1 | 5 | 1184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
