siRNA Details
Gene Symbol: Ppp1r18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126287.2 | NP_001119759.2 | 2 | 520 | ||
XM_006255952.2 | XP_006256014.1 | 1 | 2244 | |||
XM_006255953.2 | XP_006256015.1 | 2 | 1312 | |||
GenBank mRNA | BC161960.1 | - | 2 | 520 | 9027646 |
