siRNA Details
Gene Symbol: Ltb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212507.2 | NP_997672.1 | 2 | 367 | ||
GenBank mRNA | BC088208.1 | - | 2 | 367 | 7381953 | |
FQ227971.1 | - | 2 | 321 | |||
FQ235217.1 | - | 2 | 292 |
Gene Symbol: LOC103694381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772776.1 | XP_008770998.1 | 2 | 662 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
