siRNA Details
Gene Symbol: Rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025727.1 | NP_001020898.1 | 2 | 232 | ||
XM_006256225.2 | XP_006256287.1 | 2 | 309 | |||
XM_006256226.2 | XP_006256288.1 | 2 | 312 | |||
GenBank mRNA | BC099079.1 | - | 2 | 232 | 7443362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
