siRNA Details
Gene Symbol: Bicc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108531.1 | NP_001102001.1 | 9 | 1008 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_031397, AK089066, BC062174, BC111523, AF319464, and others.
Gene Ontology Categories:
