siRNA Details
Gene Symbol: RGD1306739 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134576.1 | NP_001128048.1 | 4 | 472 | ||
XM_006256355.2 | XP_006256417.1 | 4 | 503 | |||
XM_006256356.2 | XP_006256418.1 | 4 | 478 | |||
XM_008772887.1 | XP_008771109.1 | 4 | 536 |
