siRNA Details
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108532.1 | NP_001102002.1 | 171 | |||
XM_006256376.2 | XP_006256438.1 | 3 | 1826 | |||
XM_006256377.2 | XP_006256439.1 | 3 | 1765 | |||
XM_006256378.2 | XP_006256440.1 | 3 | 1745 | |||
XM_006256379.2 | XP_006256441.1 | 3 | 2405 | |||
XM_006256380.2 | XP_006256442.1 | 2 | 558 | |||
GenBank mRNA | FQ215209.1 | - | 3 | 395 |
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751958.3 | XP_003752006.2 | 2 | 565 | ||
XM_006256396.2 | XP_006256458.1 | 2 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
