siRNA Details
Gene Symbol: LOC100910795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751958.3 | XP_003752006.2 | 5 | 986 | ||
XM_006256396.2 | XP_006256458.1 | 5 | 1031 |
Gene Symbol: Hnrnph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108532.1 | NP_001102002.1 | 637 | |||
XM_006256376.2 | XP_006256438.1 | 6 | 2292 | |||
XM_006256377.2 | XP_006256439.1 | 6 | 2231 | |||
XM_006256378.2 | XP_006256440.1 | 6 | 2211 | |||
XM_006256379.2 | XP_006256441.1 | 6 | 2871 | |||
XM_006256380.2 | XP_006256442.1 | 5 | 979 | |||
GenBank mRNA | FQ215209.1 | - | 6 | 861 |

More Information
Other Species Targeted

Gene Ontology Categories:
