siRNA Details
Gene Symbol: Spock2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108533.2 | NP_001102003.1 | 1 | 155 | ||
XM_006256419.2 | XP_006256481.1 | 1 | 802 |

More Information
Other Species Targeted

Gene Ontology Categories:
