siRNA Details
Gene Symbol: Eif4ebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033069.1 | NP_001028241.1 | 2 | 417 | ||
XM_008772891.1 | XP_008771113.1 | 2 | 355 | |||
GenBank mRNA | BC100638.1 | - | 2 | 417 | 5600115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
