siRNA Details
Gene Symbol: Vps26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007740.1 | NP_001007741.1 | 6 | 711 | ||
XM_006256460.2 | XP_006256522.1 | 6 | 834 | |||
XM_008772892.1 | XP_008771114.1 | 6 | 839 | |||
GenBank mRNA | BC079150.1 | - | 6 | 711 | 7128580 | |
FQ211877.1 | - | 6 | 724 |
