siRNA Details
Gene Symbol: Tube1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108536.1 | NP_001102006.1 | 8 | 1045 | ||
XM_006256528.2 | XP_006256590.1 | 9 | 963 | |||
XM_006256529.1 | XP_006256591.1 | 8 | 833 | |||
GenBank mRNA | BC158720.1 | - | 9 | 912 |
