siRNA Details
Gene Symbol: Traf3ip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044248.1 | NP_001037713.1 | 7, 8 | 1562 | ||
XM_006256532.2 | XP_006256594.1 | 7, 8 | 1779 | |||
XM_006256533.2 | XP_006256595.1 | 6, 7 | 1274 | |||
GenBank mRNA | BC100099.1 | - | 7, 8 | 1562 | 7376204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
