siRNA Details
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108538.1 | NP_001102008.1 | 10 | 1305 | ||
XM_008773000.1 | XP_008771222.1 | 12 | 1275 | |||
GenBank mRNA | BC166724.1 | - | 12 | 1236 | 7933637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
