siRNA Details
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012177.1 | NP_001012177.1 | 14 | 1370 | ||
XM_008760678.1 | XP_008758900.1 | 13 | 1442 | |||
GenBank mRNA | BC083581.1 | - | 14 | 1370 | 7129446 |
